Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis)
Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis)
Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis)

Form: Approx.0.97g/slug | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Pc.
5 Pc.
25 Pc.
Catalog number 013394.KG
also known as 013394-KG
Price (EUR)
291,00
Each
Quantity:
1 Pc.
Request bulk or custom format
Price (EUR)
291,00
Each
Specifications
AppearanceGold colored slug
Assay Percent Range99.99% (Metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold
View more
Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only