Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 013404.A1
1 kg, Each
Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 013404.A1
1 kg, Each
Quantity
Catalog number: 013404.A1
also known as 013404-A1
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
10377-48-7
IUPAC Name
dilithium(1+) sulfate
Molecular Formula
Li2O4S
InChI Key
INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Loss on Drying
1.0% max
Total Metal Impurities
0.3% max.

Description

Lithium sulfate is used in the treatment of bipolar disorder. It acts as an important component of ion conducting glasses, solar panels and in battery. It serves as an additive for Portland cement. Further, it is used in the production of special high-strength glass, food and flavoring agent in synthetic beverages. In addition to this, it is used as a catalyst in organic synthesis for the preparation of 1-butene from n-butyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H319 - Causes serious eye irritation

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P337 + P313 - If eye irritation persists: Get medical advice/attention