Thermo Scientific Chemicals

Sodium phosphate, tribasic, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 013438.A1
1 kg, Each
Thermo Scientific Chemicals

Sodium phosphate, tribasic, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 013438.A1
1 kg, Each
Quantity
Catalog number: 013438.A1
also known as 013438-A1
Price (USD)
Price: 58.80
Online price: 49.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7601-54-9
IUPAC Name
trisodium phosphate
Molecular Formula
Na3O4P
InChI Key
RYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%

Description

Sodium phosphate, tribasic is used in soaps, detergents, photographic developers, water softeners and boiler scale removers. It is used as a food additive, as cleaning agents, as a protein precipitant and an active ingredient in fluxes designed to deoxygenate nonferrous metals for casting. It is used to prepare chlorinated trisodium phosphate by reacting with sodium hypochlorite and used as a disinfectant and bleach. Further, it is used in dairy substitutes and to inhibit mold. In addition, it is employed in ceramic production. It is also used for the preservation of biologically active soil samples and fractionization of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text