Thermo Scientific Chemicals

Sodium sulfite, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 013454.36
500 g, Each
Thermo Scientific Chemicals

Sodium sulfite, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 013454.36
500 g, Each
Quantity
Catalog number: 013454.36
also known as 013454-36
Price (USD)
Price: 55.60
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7757-83-7
IUPAC Name
disodium heptahydrate sulfite
Molecular Formula
H14Na2O10S
InChI Key
WMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
Impurity content
Cl 0.02%
Impurity content
Fe 0.001%
Impurity content
Free acid P.T.
Insoluble material
0.005%
Trace Metals
(as Pb) 0.001%

Description

Sodium sulfite is used in the photography industry to protect developing solutions from oxidation; in the pulp and paper industry; in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in the textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and manufacture of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text