Thermo Scientific Chemicals

Terbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013593.09
10 g, Each
Thermo Scientific Chemicals

Terbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013593.09
10 g, Each
Quantity
Catalog number: 013593.09
also known as 013593-09
Price (USD)
Price: 189.00
Online price: 160.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10043-27-3
IUPAC Name
terbium(3+) trinitrate
Molecular Formula
N3O9Tb
InChI Key
YJVUGDIORBKPLC-UHFFFAOYSA-N
SMILES
[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Terbium(III) nitrate hydrate is used in ceramics, glass, phosphors, lasers, and also is the important dopant for fibre amplifiers. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text