Thermo Scientific Chemicals

Cadmium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013667.14
25 g, Each
Thermo Scientific Chemicals

Cadmium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013667.14
25 g, Each
Quantity
Catalog number: 013667.14
also known as 013667-14
Price (USD)

Chemical Identifiers

CAS
10108-64-2
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Residual water
< 100 ppm
Total Metal Impurities
0.002% max.
Particle Size
-10 Mesh
Appearance (Color)
White
Form
Beads

Description

Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text