Thermo Scientific Chemicals

Potassium iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013676.06
5 g, Each
Thermo Scientific Chemicals

Potassium iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 013676.06
5 g, Each
Quantity
Catalog number: 013676.06
also known as 013676-06
Price (USD)
Price: 63.00
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7681-11-0
IUPAC Name
potassium iodide
Molecular Formula
IK
InChI Key
NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES
[K+].[I-]
Form
Beads
Residual water
≤100ppm
Total Metal Impurities
≤0.002% (≤20ppm)
Particle Size
-10 Mesh
Appearance (Color)
White

Description

Potassium iodide is a simple iodide salt utilized as a fluorescence quenching agent. It is used in photography and as an analytical reagent, It is used in treating overactive thyroid and protecting the thyroid gland from the effects of radiation from inhaled or swallowed radioactive iodine. It is used as a nutrition supplement to prevent goitre and other iodine deficiency in human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text