Thermo Scientific Chemicals

Aluminum ammonium sulfate dodecahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013802.A3
2 kg, Each
Thermo Scientific Chemicals

Aluminum ammonium sulfate dodecahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013802.A3
2 kg, Each
Quantity
Catalog number: 013802.A3
also known as 013802-A3
Price (USD)
Price: 238.00
Online price: 202.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7784-26-1
IUPAC Name
aluminium(3+) ammonium dodecahydrate disulfate
Molecular Formula
AlH28NO20S2
InChI Key
WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
White
Form
powder or crystals
Assay (unspecified)
≥99.0%

Description

Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper platingAluminum ammonium sulfate dodecahydrate acts as a crystalline double sulfate. It is used in water purification, in vegetable glues, in porcelain cements, in deodorant, in tanning and in fireproofing textiles. It serves as an ingredient in animal repellant sprays. Further, it is used in the preparation of Alum (Al(OH)3) gel. In addition, it is used in cosmetic formulation, fertilizers, food preservation and in the heating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text