Nickel Chromium wire, 0.143mm (0.0056in) dia
Nickel Chromium wire, 0.143mm (0.0056in) dia
Nickel Chromium wire, 0.143mm (0.0056in) dia
Thermo Scientific Chemicals

Nickel Chromium wire, 0.143mm (0.0056in) dia

High resistivity material for electronics industry | CAS: 11106-97-1 | CrNi | 110.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
500 m
Catalog number 013966.H4
also known as 013966-H4
Price (EUR)
36,20
Each
Quantity:
100 m
Request bulk or custom format
Price (EUR)
36,20
Each
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment0.143 mm diameter
CommentNi 80% min
High resistivity material for electronics industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only