Nickel Chromium wire, 0.51mm (0.02in) dia
Nickel Chromium wire, 0.51mm (0.02in) dia
Nickel Chromium wire, 0.51mm (0.02in) dia
Thermo Scientific Chemicals

Nickel Chromium wire, 0.51mm (0.02in) dia

CAS: 11106-97-1 | CrNi | 110.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
500 m
Catalog number 013967.H4
also known as 013967-H4
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Quantity:
100 m
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.51 mm diameter
Assay from Suppliers CofAChromium: 17.5-22.5%
Assay from Suppliers CofANickel: 77.5-82.5%
These alloys are typically used to increase the resistivity of wire. They are also used in some dental restorations (fillings). It is commonly used in ceramic as an internal support. Nichrome wire is used for its ability to withstand the high temperatures that occur when clay work is fired in a kiln.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only