Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
Thermo Scientific Chemicals

Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
50 g
500 g
2 kg
Catalog number 013990.A9
also known as 013990-A9
Price (USD)
399.65
Special offer
Online exclusive
470.00
Save 70.35 (15%)
Each
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (USD)
399.65
Special offer
Online exclusive
470.00
Save 70.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only