Thermo Scientific Chemicals

Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals

Catalog number: 014051.14
25 g, Each
Thermo Scientific Chemicals

Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals

Catalog number: 014051.14
25 g, Each
Quantity
Catalog number: 014051.14
also known as 014051-14
Price (USD)
Price: 284.00
Online price: 241.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-95-0
IUPAC Name
λ²-tin(2+) sulfanediide
Molecular Formula
SSn
InChI Key
DZXKSFDSPBRJPS-UHFFFAOYSA-N
SMILES
[S--].[Sn++]
Assay (unspecified)
99.5% min

Description

Polymerization catalyst.Tin(II) sulfide is used as a polymerization catalyst. It is also used as a friction material. Further, it is used in optoelectronic device as well as a component in heterojunction photovoltaic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text