Thermo Scientific Chemicals

Sodium carbonate monohydrate, 85% min, Thermo Scientific Chemicals

Catalog number: 014108.A7
5 kg, Each
Thermo Scientific Chemicals

Sodium carbonate monohydrate, 85% min, Thermo Scientific Chemicals

Catalog number: 014108.A7
5 kg, Each
Quantity
Catalog number: 014108.A7
also known as 014108-A7
Price (USD)
Price: 88.10
Online price: 74.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5968-11-6
IUPAC Name
disodium hydrate carbonate
Molecular Formula
CH2Na2O4
InChI Key
MQRJBSHKWOFOGF-UHFFFAOYSA-L
SMILES
O.[Na+].[Na+].[O-]C([O-])=O
Assay (unspecified)
85% min

Description

Sodium carbonate monohydrate is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text