Óxido de di-n-butilestaño, Thermo Scientific Chemicals
Óxido de di-n-butilestaño, Thermo Scientific Chemicals
Óxido de di-n-butilestaño, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Óxido de di-n-butilestaño, Thermo Scientific Chemicals

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
50 g
1 kg
Número de catálogo 014117.A1
también denominado 014117-A1
Precio (EUR)
189,00
Each
Cantidad:
1 kg
Pedido a granel o personalizado
Precio (EUR)
189,00
Each
Especificaciones
Beilstein4126243
CAS818-08-6
Nombre del producto químico o materialDi-n-butyltin oxide
ColorBlanco
Información de PUNTOSClase de peligro de transporte:6,1; grupo de embalaje:II; Nombre del producto expedido:COMPUESTOS DE ORGANOTINA, SÓLIDO, SIN ESPECIFICAR
Ver más
Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only