Nickel(II) fluoride tetrahydrate, 98+%
Nickel(II) fluoride tetrahydrate, 98+%
Nickel(II) fluoride tetrahydrate, 98+%
Thermo Scientific Chemicals

Nickel(II) fluoride tetrahydrate, 98+%

CAS: 13940-83-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 014132.30
also known as 014132-30
Price (EUR)
161,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
161,00
Each
Chemical Identifiers
CAS13940-83-5
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.1% min
Nickel(II) fluoride tetrahydrate is used in surface treatment of aluminum profile and printing ink making, fluorescence lamp, fluidizer and catalyzer for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only