Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)
Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)
Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)
Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)
Thermo Scientific Chemicals

Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 014153.30
also known as 014153-30
Price (USD)
1,790.65
Special offer
Online exclusive
2,106.00
Save 315.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,790.65
Special offer
Online exclusive
2,106.00
Save 315.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Assay from Suppliers CofATotal metal impurities: ≤0.001% (≤10ppm)
Diameter90% is ≥1.5mm to ≤3.5mm
FormShot
For vapor deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only