Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Thermo Scientific Chemicals

Potassium, Oil based standard solution, Specpure™ K 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014216.18
also known as 014216-18
50 g
Catalog number 014216.18
also known as 014216-18
Price (EUR)
95,80
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
95,80
Each
Specifications
Chemical Name or MaterialPotassium Oil based standard solution
MDL NumberMFCD00133776
Name NoteK 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only