Bismuth, Oil based standard solution, Specpure™ Bi 1000μg/g
Bismuth, Oil based standard solution, Specpure™ Bi 1000μg/g
Bismuth, Oil based standard solution, Specpure™ Bi 1000μg/g
Thermo Scientific Chemicals

Bismuth, Oil based standard solution, Specpure™ Bi 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014223.18
also known as 014223-18
50 g
Catalog number 014223.18
also known as 014223-18
Price (TWD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth Oil based standard solution
MDL NumberMFCD00134033
Name NoteBi 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only