Calcium, Oil based standard solution, Specpure™ Ca 1000μg/g
Thermo Scientific Chemicals

Calcium, Oil based standard solution, Specpure™ Ca 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014225.18
also known as 014225-18
50 g
Catalog number 014225.18
also known as 014225-18
Price (EUR)
81,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
81,00
Each
Specifications
Chemical Name or MaterialCalcium Oil based standard solution
MDL NumberMFCD00085314
Name NoteCa 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only