Thermo Scientific Chemicals

Potassium nitrite, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog number: 014245.22
100 g, Each
Thermo Scientific Chemicals

Potassium nitrite, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog number: 014245.22
100 g, Each
Quantity
Catalog number: 014245.22
also known as 014245-22
Price (USD)

Chemical Identifiers

CAS
7758-09-0
IUPAC Name
potassium nitrite
Molecular Formula
KNO2
InChI Key
BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES
[K+].[O-]N=O
Impurity content
Na 0.5%
Impurity content
SO₄ 0.01%
Insoluble material
0.01%
Trace Metals
(as Pb) 0.001%
Assay (unspecified)
96.0% KNO₂ min

Description

In analytical chemistry (testing for amino acids, cobalt, iodine, urea).Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text