Iodine, crystalline, 99.5%, Thermo Scientific Chemicals
Iodine, crystalline, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodine, crystalline, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
100 g
200 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014248.A1
also known as 014248-A1
Price (USD)/ Each
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Add to cart
Chemical Identifiers
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
Molecular Weight (g/mol)253.81
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.50%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only