Thermo Scientific Chemicals

Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog number: 014365.14
25 g, Each
Thermo Scientific Chemicals

Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog number: 014365.14
25 g, Each
Quantity
Catalog number: 014365.14
also known as 014365-14
Price (USD)

Chemical Identifiers

CAS
1306-25-8
Total Metal Impurities
0.001% max.

Description

Cadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text