Nickel, plasma standard solution, Specpure™ Ni 10,000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 10,000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 10,000μg/mL
Thermo Scientific Chemicals

Nickel, plasma standard solution, Specpure™ Ni 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014414.AD
also known as 014414-AD
Price (EUR)
94,20
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
94,20
Each
Specifications
Chemical Name or MaterialNickel plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H314-H317-H335-H350-H373
Health Hazard 2GHS H Statement
H314-H351-H373-H317
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
View more
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only