Dysprosium, plasma standard solution, Specpure™ Dy 10,000μg/mL
Dysprosium, plasma standard solution, Specpure™ Dy 10,000μg/mL
Thermo Scientific Chemicals

Dysprosium, plasma standard solution, Specpure™ Dy 10,000μg/mL

Have Questions?
變更視圖buttonViewtableView
Quantity:
50 mL
100 mL
500 mL
產品號碼 014426.AP
亦稱為 014426-AP
價格 (TWD)
-
申請報價
Quantity:
500 mL
Request bulk or custom format
規格
Chemical Name or MaterialDysprosium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only