Cesium nitrate, 99.99% (metals basis)
Cesium nitrate, 99.99% (metals basis)
Cesium nitrate, 99.99% (metals basis)
Thermo Scientific Chemicals

Cesium nitrate, 99.99% (metals basis)

CAS: 7789-18-6 | CsNO3 | 194.909 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 014440.14
also known as 014440-14
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Chemical Identifiers
CAS7789-18-6
IUPAC Namecaesium(1+) nitrate
Molecular FormulaCsNO3
InChI KeyNLSCHDZTHVNDCP-UHFFFAOYSA-N
SMILES[Cs+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only