Thermo Scientific Chemicals

Barium carbonate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 014452.09
10 g, Each
Thermo Scientific Chemicals

Barium carbonate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 014452.09
10 g, Each
Quantity
Catalog number: 014452.09
also known as 014452-09
Price (USD)
Quantity
-

Chemical Identifiers

CAS
513-77-9
IUPAC Name
barium(2+) carbonate
Molecular Formula
CBaO3
InChI Key
AYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES
[Ba++].[O-]C([O-])=O
Appearance (Color)
White
Form
powder and/or chunks
Assay (special)
?2.5 % of Theoretical value (Ba)
Total Metal Impurities
≤0.05% (500 ppm)

Description

Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text