Thermo Scientific Chemicals

Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014485.14
25 g, Each
Thermo Scientific Chemicals

Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014485.14
25 g, Each
Quantity
Catalog number: 014485.14
also known as 014485-14
Price (USD)
Price: 35.60
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7803-63-6
IUPAC Name
ammonium hydrogen sulfate
Molecular Formula
H5NO4S
InChI Key
BIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES
[NH4+].OS([O-])(=O)=O
Total Metal Impurities
0.1% max.

Description

Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text