Sodium nitrate, ACS, 99.0% min
Sodium nitrate, ACS, 99.0% min
Sodium nitrate, ACS, 99.0% min
Sodium nitrate, ACS, 99.0% min
Thermo Scientific Chemicals

Sodium nitrate, ACS, 99.0% min

CAS: 7631-99-4 | NNaO3 | 84.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014493.A1
also known as 014493-A1
Price (EUR)
66,65
Online Exclusive
74,10
Save 7,45 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
66,65
Online Exclusive
74,10
Save 7,45 (10%)
Each
Chemical Identifiers
CAS7631-99-4
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% NaNO₃ min
Impurity contentCl 0.001%
Impurity contentFe 3ppm
Impurity contentIO₃ 5ppm
Impurity contentMg 0.002%
View more
In manufacturing of HNO{3}, as a catalyst to manufacture H{2}SO{4}, in manufacturing of glass, enamel for pottery, sodium nitriteSodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only