Thermo Scientific Chemicals

Mercury(II) nitrate hydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 014497.18
50 g, Each
Thermo Scientific Chemicals

Mercury(II) nitrate hydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 014497.18
50 g, Each
Quantity
Catalog number: 014497.18
also known as 014497-18
Price (USD)

Chemical Identifiers

CAS
7783-34-8
IUPAC Name
mercury(2+) hydrate dinitrate
Molecular Formula
H2HgN2O7
InChI Key
KVICROHOONHSRH-UHFFFAOYSA-N
SMILES
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Comment
Residue after reduction 0.01%
Impurity content
Cl 0.002%
Impurity content
Fe 0.001%
Impurity content
SO₄ 0.002%
Assay (unspecified)
98.0% HgN₂O₆.xH₂O (x=₁-₂) min

Description

Mercury(II) nitrate is used as an oxidizing agent involved in organic synthesis. Its reaction with acetone yielded an organometallic compound containing mercury. In addition, it serves as a nitrification agent and analytical reagents used in the laboratory. It is involved in the preparation of mercury fulminate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text