Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Holmium(III) acetate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Holmium(III) acetate hydrate, REacton™, 99.9% (REO)

CAS: 312619-49-1 | C6H9HoO6 | 342.06 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
50 g
產品號碼 014591.09
亦稱為 014591-09
價格 (TWD)
-
Quantity:
10 g
Request bulk or custom format
化學識別
CAS312619-49-1
IUPAC Nameholmium(3+) triacetate
Molecular FormulaC6H9HoO6
InChI KeyHWQRVFRWLQGBFV-UHFFFAOYSA-K
SMILES[Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
檢視更多
規格Specification Sheet規格表
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only