Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO)

CAS: 13842-67-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 014596.18
also known as 014596-18
Price (USD)
793.65
Online Exclusive
882.00
Save 88.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
793.65
Online Exclusive
882.00
Save 88.35 (10%)
Each
Chemical Identifiers
CAS13842-67-6
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only