Praseodymium(III) sulfate octahydrate, 99.9% (REO)
Praseodymium(III) sulfate octahydrate, 99.9% (REO)
Praseodymium(III) sulfate octahydrate, 99.9% (REO)
Thermo Scientific Chemicals

Praseodymium(III) sulfate octahydrate, 99.9% (REO)

CAS: 13510-41-3 | H16O20Pr2S3 | 714.103 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 014607.14
also known as 014607-14
Price (EUR)
60,30
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
60,30
Each
Chemical Identifiers
CAS13510-41-3
IUPAC Namedipraseodymium(3+) octahydrate trisulfate
Molecular FormulaH16O20Pr2S3
InChI KeyNIJXLQIGVGZNKP-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Pr+3].[Pr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only