Thermo Scientific Chemicals

Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals

Catalog number: 014660.36
500 g, Each
Thermo Scientific Chemicals

Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals

Catalog number: 014660.36
500 g, Each
Quantity

Chemical Identifiers

CAS
1341-49-7
IUPAC Name
amine dihydrofluoride
Molecular Formula
F2H5N
InChI Key
KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES
N.F.F
Assay (unspecified)
95% min

Description

Ammonium hydrogen fluoride is used as a chemical reagent in analysis and an antiseptic. It is also used in ceramics, electroplating, component of glass etchants as well as food processing equipment disinfectant. It acts as an intermediate in the production of hydrofluoric acid from hexafluorosilicic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text