Copper(II) gluconate
Copper(II) gluconate
Copper(II) gluconate
Thermo Scientific Chemicals

Copper(II) gluconate

CAS: 527-09-3 | C12H22CuO14 | 453.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 015325.30
also known as 015325-30
Price (USD)
112.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS527-09-3
IUPAC Namecopper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular FormulaC12H22CuO14
InChI KeyOCUCCJIRFHNWBP-IYEMJOQQSA-L
SMILES[Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to blue-green
FormPowder
Assay from Supplier's CofA≥97.5%
Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only