Thermo Scientific Chemicals

Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 015546.A5
3 kg, Each
Thermo Scientific Chemicals

Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 015546.A5
3 kg, Each
Quantity
Catalog number: 015546.A5
also known as 015546-A5
Price (USD)
Quantity
-

Chemical Identifiers

CAS
79-41-4
IUPAC Name
2-methylprop-2-enoic acid
Molecular Formula
C4H6O2
InChI Key
CERQOIWHTDAKMF-UHFFFAOYSA-N
SMILES
CC(=C)C(O)=O
Stabilizer
MEHQ (180 to 275 ppm)
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%

Description

Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly (methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text