Thermo Scientific Chemicals

Pentaerythritol triacrylate, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 015705.36
500 g, Each
Thermo Scientific Chemicals

Pentaerythritol triacrylate, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 015705.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 015705.36
also known as 015705-36
Price (USD)
Price: 68.50
Online price: 58.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3524-68-3
IUPAC Name
3-hydroxy-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate
Molecular Formula
C14H18O7
InChI Key
HVVWZTWDBSEWIH-UHFFFAOYSA-N
SMILES
OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
Form
Liquid
Stabilizer
MEHQ
Appearance (Color)
Clear to cloudy, colorless to pale yellow
Viscosity
≤800 CPS (25°C)
Assay from Suppliers CofA
≥97.0%

Description

Pentaerythritol triacrylate is used in UV-cured inks, dental polymers and varnishes. Further, it is used in the preparation of printing inks, coatings, in photoresists, acrylic glues, adhesives, binders, paper and wood impregnates. In addition, it serves as a modifier for polyester and fiberglass and plastic components for the medical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text