Thermo Scientific Chemicals

Beryllium sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 016104.14
25 g, Each
Thermo Scientific Chemicals

Beryllium sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 016104.14
25 g, Each
Quantity

Chemical Identifiers

CAS
7787-56-6
IUPAC Name
beryllium(2+) tetrahydrate sulfate
Molecular Formula
BeH8O8S
InChI Key
DIMYTQPLZWDZFE-UHFFFAOYSA-L
SMILES
[Be++].O.O.O.O.[O-]S([O-])(=O)=O
Total Metal Impurities
0.01% max.

Description

In Industries it is used for beryllium salt making, ceramics and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text