Thermo Scientific Chemicals

Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max, Thermo Scientific Chemicals

Catalog number: 016791.18
50 g, Each
Thermo Scientific Chemicals

Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max, Thermo Scientific Chemicals

Catalog number: 016791.18
50 g, Each
Quantity
Catalog number: 016791.18
also known as 016791-18
Price (USD)
Price: 111.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10476-81-0
Assay (unspecified)
1% max.
Impurity content
Ba 0.1% max

Description

Used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text