Thermo Scientific Chemicals

Acetone-d{6}, 99.9% (Isotopic), Thermo Scientific Chemicals

Catalog number: 016797.50
5 x 10 g, Each
Thermo Scientific Chemicals

Acetone-d{6}, 99.9% (Isotopic), Thermo Scientific Chemicals

Catalog number: 016797.50
5 x 10 g, Each
Quantity
Catalog number: 016797.50
also known as 016797-50
Price (USD)
Have Questions?

Chemical Identifiers

CAS
666-52-4
IUPAC Name
(²H₆)propan-2-one
Molecular Formula
C3H6O
InChI Key
CSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
Form
liquid
Proton NMR
>/= 99.9% ( Isotopic purity)
Assay (unspecified)
99.5% min.- ( chemical purity by NMR)
Color Change
Clear, colorless

Description

Acetone-d{6} is employed as an NMR solvent. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text