Bis(cyclopentadienyl)cobalt
Bis(cyclopentadienyl)cobalt
Bis(cyclopentadienyl)cobalt
Thermo Scientific Chemicals

Bis(cyclopentadienyl)cobalt

CAS: 1277-43-6 | C10H10Co | 189.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 016857.14
also known as 016857-14
Price (USD)
857.65
Online Exclusive
953.00
Save 95.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
857.65
Online Exclusive
953.00
Save 95.35 (10%)
Each
Chemical Identifiers
CAS1277-43-6
IUPAC Nameλ²-cobalt(2+) bis(cyclopenta-2,4-dien-1-ide)
Molecular FormulaC10H10Co
InChI KeyILZSSCVGGYJLOG-UHFFFAOYSA-N
SMILES[Co++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder and /or chunks
Elemental AnalysisC ≥62.2%
Appearance (Color)Purple to black
Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only