4,4-Dimethyl-2-cyclohexen-1-one, 96%
4,4-Dimethyl-2-cyclohexen-1-one, 96%
4,4-Dimethyl-2-cyclohexen-1-one, 96%
Thermo Scientific Chemicals

4,4-Dimethyl-2-cyclohexen-1-one, 96%

CAS: 1073-13-8 | C8H12O | 124.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 017162.06
also known as 017162-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1073-13-8
IUPAC Name4,4-dimethylcyclohex-2-en-1-one
Molecular FormulaC8H12O
InChI KeyHAUNPYVLVAIUOO-UHFFFAOYSA-N
SMILESCC1(C)CCC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥95.0% (non-U.S. specification)
Assay from Suppliers CofA≥95.0% (GC, U.S. specification)
CommentPurchased in the U.S. and in other countries
Refractive Index1.4710-1.4750 @ 20°C (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only