2,4,6-Triisopropylbenzenesulfonyl hydrazide
2,4,6-Triisopropylbenzenesulfonyl hydrazide
2,4,6-Triisopropylbenzenesulfonyl hydrazide
2,4,6-Triisopropylbenzenesulfonyl hydrazide
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzenesulfonyl hydrazide

As a reagent for generation of diimide | CAS: 39085-59-1 | C15H26N2O2S | 298.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 017976.02
also known as 017976-02
Price (EUR)
39,30
Each
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
39,30
Each
Add to cart
Specifications
Beilstein2145001
CAS39085-59-1
Chemical Name or Material2, 4, 6-Triisopropylbenzenesulfonyl hydrazide
EINECS Number254-282-7
Formula Weight298.58
View more
As a reagent for generation of diimide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only