Di-n-butyltin diacetate, 95%
Di-n-butyltin diacetate, 95%
Di-n-butyltin diacetate, 95%
Thermo Scientific Chemicals

Di-n-butyltin diacetate, 95%

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions | CAS: 1067-33-0 | C12H26O4Sn | 353.05 g/mol
数量:
50 g
250 g
1 kg
製品番号(カタログ番号) 018595.A1
または、製品番号018595-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1067-33-0
Molecular FormulaC12H26O4Sn
InChI KeyLDZDQDDROJXUBO-UHFFFAOYSA-N
SMILESCC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
Molecular Weight (g/mol)353.05
Assay (Titration)32.2 to 34.2 wt% total Sn
Appearance (Color)Colorless to pale yellow to pale brown
FormLiquid
Assay (unspecified)33.6 to 35.6 wt% acetic acid
As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only