Scandium(III) chloride, anhydrous, 99.9% (REO)
Scandium(III) chloride, anhydrous, 99.9% (REO)
Scandium(III) chloride, anhydrous, 99.9% (REO)
Thermo Scientific Chemicals

Scandium(III) chloride, anhydrous, 99.9% (REO)

CAS: 10361-84-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 018677.01
also known as 018677-01
Price (USD)
76.65
Online Exclusive
85.10
Save 8.45 (10%)
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
85.10
Save 8.45 (10%)
Each
Chemical Identifiers
CAS10361-84-9
IUPAC Namescandium(3+) trichloride
Molecular FormulaCl3Sc
InChI KeyDVMZCYSFPFUKKE-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sc+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Scandium(III) chloride is a Prins cyclization catalyst. Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only