Copper(II) methoxide, typically 98% (metals basis)
Thermo Scientific Chemicals

Copper(II) methoxide, typically 98% (metals basis)

CAS: 1184-54-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 018861.06
also known as 018861-06
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS1184-54-9
CAS1184-54-9
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤2.0%
Appearance (Color)Blue to green
FormPowder or crystals or crystalline powder
Appearance (Color)Blue to green
FormPowder or crystals or crystalline powder
View more
Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only