Tetra-n-butylammonium acetate
Tetra-n-butylammonium acetate
Tetra-n-butylammonium acetate
Tetra-n-butylammonium acetate
Thermo Scientific Chemicals

Tetra-n-butylammonium acetate

CAS: 10534-59-5 | C18H39NO2 | 301.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 019279.03
also known as 019279-03
Price (EUR)
37,30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
37,30
Each
Add to cart
Chemical Identifiers
CAS10534-59-5
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder
Appearance (Color)White to yellow
Assay (Titration)>95.0%
Tetra-n-butylammonium acetate  is used as catalyst, curing agent, and initiator in the polymer industry. It is also used catalyst for the alkynylation of carbonyl compounds with trimethylsilylacetylenes. It is also used as PE/PP clarifying agent, epoxy polymerization initiator, surfactants, emulsifiers, and antistatic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only