Thermo Scientific Chemicals

Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals

Catalog number: 019347.22
100 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals

Catalog number: 019347.22
100 g, Each
Quantity
Catalog number: 019347.22
also known as 019347-22
Price (USD)
Price: 207.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
37451-68-6
IUPAC Name
tetrabutylazanium chloride
Molecular Formula
C16H36ClN
InChI Key
NHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES
[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
Assay (Titration)
≥98.0 to ≤102.0% (as 1:1 solvate with ethanol)
Form
Crystals or powder or crystalline powder or chunks
Impurity content
Tributylamine Hydrochloride: ≤0.2%
Appearance (Color)
White to pale yellow

Description

Tetra-n-butylammonium chloride hydrate is used with phosphorus pentoxide for greener deoxychlorination. It is used as a phase transfer catalyst for the preparation of 2-(3-pyridyl)-1-methylpyrrole by the reaction between 3-bromopyridine and 1-methyl-2pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text