Toluene, Spectrophotometric Grade, 99.7+%
Toluene, Spectrophotometric Grade, 99.7+%
Toluene, Spectrophotometric Grade, 99.7+%
Thermo Scientific Chemicals

Toluene, Spectrophotometric Grade, 99.7+%

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03% | CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 019376.M6
亦稱為 019376-M6
價格 (TWD)
-
Quantity:
4 x 4 L
Request bulk or custom format
化學識別
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear, colorless
Color (APHA)10, max
Optical Absorbance at285nm, 1.00, max
Optical Absorbance at286nm, 0.70, max
Optical Absorbance at293nm, 0.20, max
檢視更多
Toluene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only