Tetra-n-propylammonium chloride, 99+%
Tetra-n-propylammonium chloride, 99+%
Tetra-n-propylammonium chloride, 99+%
Tetra-n-propylammonium chloride, 99+%
Thermo Scientific Chemicals

Tetra-n-propylammonium chloride, 99+%

CAS: 5810-42-4 | C12H28ClN | 221.813 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 019379.14
also known as 019379-14
Price (EUR)
118,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
118,00
Each
Add to cart
Chemical Identifiers
CAS5810-42-4
IUPAC Nametetrapropylazanium chloride
Molecular FormulaC12H28ClN
InChI KeyFBEVECUEMUUFKM-UHFFFAOYSA-M
SMILES[Cl-].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Assay (HPLC)≥99.0%
Identification (FTIR)Conforms
Melting point235-242°C
Tetra-n-propylammonium chloride is used as phase-transfer type applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only