Eutech™ pH Buffers and Solutions, Color-Coded
Actual product may vary
Thermo Scientific™

Eutech™ pH Buffers and Solutions, Color-Coded

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitypH
01X60810160 mL10.01
ECBU4BTC480 mL4.01
ECPHBUFKITC480 mL Ea.4.01, 7.00, 10.01
01X60800160 mL4.01
ECBU4BTC1LIT1 L4.01
ECBU7BTC480 mL7.00
01X60790160 mL7.00
ECBU7BTC1LIT1 L7.00
ECBU10BTC480 mL10.01
ECBU10BTC1LIT1 L10.01
Catalog number 01X608101
Price (KRW)
16,000
Each
Quantity:
60 mL
pH:
10.01
Price (KRW)
16,000
Each

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.

Features:

  • Buffers and calibration solutions packed in 60mL, 480 ml or 1L durable plastic bottles – economical and convenient for laboratory use.

Buffer Color Codes:

  • Pink = pH 4.01
  • Yellow = pH 7.00
  • Blue = pH 10.01
Specifications
ColorBlue
DescriptionBuffer
For Use With (Equipment)pH Meter and Electrode
Quantity60 mL
Solution TypepH Buffer
pH10.01
Unit SizeEach